siRNA Details
| Gene Symbol: Tmem218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025464.2 | NP_079740.1 | 4 | 420 | ||
| GenBank mRNA | AK003010.1 | - | 4 | 417 | ||
| AK007575.1 | - | 4 | 404 | |||
| AK008902.1 | - | 4 | 391 | |||
| AK161115.1 | - | 422 | ||||
| BC034160.1 | - | 4 | 420 | 5049687 | ||

Tmem218
Genomic Map