siRNA Details
| Gene Symbol: Gkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025467.1 | NP_079743.1 | 3 | 224 | ||
| GenBank mRNA | AK007451.1 | - | 3 | 224 | ||
| AK007705.1 | - | 3 | 224 | |||
| AK008692.1 | - | 3 | 225 | |||
| AK008986.1 | - | 3 | 225 | |||
| AY943909.1 | - | 3 | 232 | |||
| BC099445.1 | - | 3 | 213 | 30788014 | ||

Gkn2
Genomic Map