siRNA Details
| Gene Symbol: Mptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025470.3 | NP_079746.1 | 2 | 370 | ||
| GenBank mRNA | AK007658.1 | - | 2 | 371 | ||
| AK007963.1 | - | 2 | 370 | |||
| BC024348.1 | - | 2 | 357 | 4985132 | ||
| Gene Symbol: Mptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205011.2 | NP_001191940.1 | 2 | 349 | ||

Mptx1
Genomic Map