siRNA Details
| Gene Symbol: 1810032O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037979.1 | - | 5 | 617 | ||
| BC055707.1 | - | 5 | 611 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_025472.1 | - | 617 | Chr. - - [-] on Build | ||

1810032O08Rik
Genomic Map