siRNA Details
| Gene Symbol: Isoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025478.3 | NP_079754.2 | 3 | 512 | ||
| GenBank mRNA | AK011674.1 | - | 3 | 379 | ||
| AK134794.1 | - | 3 | 512 | |||
| AK152763.1 | - | 3 | 509 | |||
| AK153257.1 | - | 3 | 509 | |||
| AK165542.1 | - | 3 | 531 | |||
| AY035213.1 | - | 3 | 512 | |||
| BC016576.1 | - | 3 | 519 | 2609919 | ||

Isoc1
Genomic Map