siRNA Details
| Gene Symbol: Gm7102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177513.1 | NP_001170984.1 | 1 | 801 | ||
| GenBank mRNA | AK163227.1 | - | 1 | 805 | ||
| Gene Symbol: Mplkip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025479.5 | NP_079755.1 | 2 | 559 | ||
| GenBank mRNA | AK011919.1 | - | 2 | 554 | ||
| AK012770.1 | - | 2 | 552 | |||
| AK030630.1 | - | 2 | 560 | |||
| AK141333.1 | - | 1562 | ||||
| BC139338.1 | - | 2 | 512 | 8862360 | ||
| BC139341.1 | - | 2 | 512 | 8862363 | ||

Gm7102
Genomic Map