siRNA Details
| Gene Symbol: Mplkip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025479.5 | NP_079755.1 | 2 | 1518 | ||
| GenBank mRNA | AK030630.1 | - | 2 | 1518 | ||
| AK041757.1 | - | 1 | 419 | |||
| AK141333.1 | - | 2521 | ||||
| BC139338.1 | - | 2 | 1471 | 8862360 | ||
| BC139341.1 | - | 2 | 1471 | 8862363 | ||

Mplkip
Genomic Map