siRNA Details
| Gene Symbol: 1700018B24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018885.1 | - | 1 | 246 | ||
| BC049578.1 | - | 1 | 260 | |||
| BC061086.1 | - | 1 | 276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700018B24Rik| Gene Symbol: 1700018B24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018885.1 | - | 1 | 246 | ||
| BC049578.1 | - | 1 | 260 | |||
| BC061086.1 | - | 1 | 276 | |||