siRNA Details
| Gene Symbol: Cenpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025495.3 | NP_079771.1 | 6, 7 | 690 | ||
| GenBank mRNA | AK006234.1 | - | 6, 7 | 727 | ||
| AK012863.1 | - | 6, 7 | 694 | |||
| BC094280.1 | - | 6, 7 | 743 | |||
| BC147207.1 | - | 6, 7 | 670 | 9056262 | ||
| BC147208.1 | - | 6, 7 | 670 | 9056260 | ||

Cenpp
Genomic Map