siRNA Details
| Gene Symbol: 1700029P11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025503.4 | NP_079779.2 | 1 | 618 | ||
| GenBank mRNA | AK006381.1 | - | 1 | 618 | ||
| AK006519.1 | - | 1 | 614 | |||
| AK007222.1 | - | 1 | 614 | |||
| BC048460.1 | - | 1 | 607 | 6770795 | ||

1700029P11Rik
Genomic Map