siRNA Details
| Gene Symbol: Chmp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025519.2 | NP_079795.1 | 5 | 1258 | ||
| GenBank mRNA | AK009273.1 | - | 5 | 1260 | ||
| AK033502.1 | - | 151 | ||||
| BC006951.1 | - | 113 | ||||
| BC115983.1 | - | 5 | 1244 | 40105850 | ||
| BC117538.1 | - | 5 | 1248 | 40105849 | ||

Chmp4c
Genomic Map