siRNA Details
| Gene Symbol: 2310011J03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025521.3 | NP_079797.1 | 2 | 567 | ||
| GenBank mRNA | AK009305.1 | - | 2 | 550 | ||
| AK046452.1 | - | 2 | 528 | |||
| AK140230.1 | - | 2 | 528 | |||
| BC023429.1 | - | 2 | 534 | 5042442 | ||

2310011J03Rik
Genomic Map