siRNA Details
| Gene Symbol: Srp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025527.3 | NP_079803.2 | 3 | 211 | ||
| GenBank mRNA | AK004118.1 | - | 3 | 213 | ||
| AK009412.1 | - | 3 | 175 | |||
| AK010760.1 | - | 3 | 210 | |||
| AK037009.1 | - | 2 | 2681 | |||
| AK135508.1 | - | 4 | 338 | |||
| AK152322.1 | - | 3 | 190 | |||
| AK162107.1 | - | 3 | 181 | |||
| BC051932.1 | - | 3 | 178 | 5710637 | ||

Srp19
Genomic Map