siRNA Details
| Gene Symbol: Slmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025531.2 | NP_079807.1 | 3 | 307 | ||
| GenBank mRNA | AK003043.1 | - | 3 | 281 | ||
| AK008542.1 | - | 3 | 267 | |||
| AK013197.1 | - | 3 | 274 | |||
| BC004775.1 | - | 2 | 105 | |||
| BC026968.1 | - | 3 | 307 | 4235395 | ||

Slmo2
Genomic Map