siRNA Details
| Gene Symbol: Prl2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025532.3 | NP_079808.1 | 3 | 381 | ||
| GenBank mRNA | AF525155.1 | - | 3 | 332 | ||
| AK005411.1 | - | 3 | 382 | |||
| AK009863.1 | - | 3 | 381 | |||
| BC100329.1 | - | 3 | 371 | 30787440 | ||
| BC111112.1 | - | 3 | 377 | 40050224 | ||
| BC138199.1 | - | 3 | 374 | 8861218 | ||

Prl2b1
Genomic Map