siRNA Details
Gene Symbol: Tsfm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025537.3 | NP_079813.1 | 3, 4 | 368 | ||
GenBank mRNA | AJ277891.1 | - | 2, 3 | 205 | ||
AK009898.1 | - | 3, 4 | 388 | |||
AK012240.1 | - | 3, 4 | 359 | |||
AK018591.1 | - | 3, 4 | 357 | |||
AK020437.1 | - | 3, 4 | 353 | |||
AK169164.1 | - | 3, 4 | 359 | |||
AK171656.1 | - | 3, 4 | 365 | |||
AK172112.1 | - | 3, 4 | 365 | |||
AK215409.1 | - | 1, 2 | 73 | |||
BC034286.1 | - | 3, 4 | 347 | |||
BC057904.1 | - | 3, 4 | 368 | 5012380 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
