siRNA Details
| Gene Symbol: Alkbh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025538.3 | NP_079814.1 | 4 | 869 | ||
| NM_027372.1 | NP_081648.1 | 3 | 695 | |||
| GenBank mRNA | AK009812.1 | - | 4 | 856 | ||
| AK010930.1 | - | 3 | 696 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alkbh7| Gene Symbol: Alkbh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025538.3 | NP_079814.1 | 4 | 869 | ||
| NM_027372.1 | NP_081648.1 | 3 | 695 | |||
| GenBank mRNA | AK009812.1 | - | 4 | 856 | ||
| AK010930.1 | - | 3 | 696 | |||