siRNA Details
Gene Symbol: 2410004B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025555.4 | NP_079831.1 | 3 | 730 | ||
GenBank mRNA | AK010378.1 | - | 3 | 730 | ||
AK019404.1 | - | 3 | 726 | |||
AK019806.1 | - | 2 | 374 | |||
AK077565.1 | - | 4 | 848 | |||
BC049577.1 | - | 4 | 684 | |||
BC115465.2 | - | 3 | 729 | 40100373 |
