siRNA Details
| Gene Symbol: Cyb5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025558.5 | NP_079834.2 | 5 | 3303 | ||
| GenBank mRNA | AK052810.1 | - | 5 | 3303 | ||
| AK088970.1 | - | 5 | 3303 | |||
| AK165543.1 | - | 5 | 3305 | |||
| AK168422.1 | - | 5 | 3293 | |||
| BC046479.1 | - | 5 | 3266 | |||
| BC046810.1 | - | 5 | 3269 | |||
| BC048147.1 | - | 5 | 3293 | |||
| BC054749.1 | - | 5 | 3324 | 6410090 | ||
| BC058812.1 | - | 5 | 3264 | 6530835 | ||
| BC062980.1 | - | 5 | 3317 | 6842939 | ||

Cyb5b
Genomic Map