siRNA Details
| Gene Symbol: Oxld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025560.2 | NP_079836.1 | 2 | 788 | ||
| GenBank mRNA | AK007842.1 | - | 2 | 789 | ||
| AK021150.1 | - | 2 | 776 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oxld1| Gene Symbol: Oxld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025560.2 | NP_079836.1 | 2 | 788 | ||
| GenBank mRNA | AK007842.1 | - | 2 | 789 | ||
| AK021150.1 | - | 2 | 776 | |||