siRNA Details
| Gene Symbol: 2010012O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025563.3 | NP_079839.1 | 5 | 1160 | ||
| GenBank mRNA | AK046524.1 | - | 5 | 1159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010012O05Rik| Gene Symbol: 2010012O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025563.3 | NP_079839.1 | 5 | 1160 | ||
| GenBank mRNA | AK046524.1 | - | 5 | 1159 | ||