siRNA Details
| Gene Symbol: Fam136a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025591.2 | NP_079867.1 | 3 | 548 | ||
| GenBank mRNA | BC024049.1 | - | 3 | 548 | 5100000 | |
| BC103775.1 | - | 3 | 549 | 6485116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam136a| Gene Symbol: Fam136a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025591.2 | NP_079867.1 | 3 | 548 | ||
| GenBank mRNA | BC024049.1 | - | 3 | 548 | 5100000 | |
| BC103775.1 | - | 3 | 549 | 6485116 | ||