siRNA Details
| Gene Symbol: Fam136a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025591.2 | NP_079867.1 | 3 | 660 | ||
| GenBank mRNA | AK005280.1 | - | 3 | 620 | ||
| AK141279.1 | - | 3 | 620 | |||
| BC013621.1 | - | 3 | 608 | |||
| BC024049.1 | - | 3 | 660 | 5100000 | ||
| BC103775.1 | - | 3 | 661 | 6485116 | ||

Fam136a
Genomic Map