siRNA Details
| Gene Symbol: Zmat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025594.3 | NP_079870.1 | 6 | 792 | ||
| GenBank mRNA | AK007521.1 | - | 1 | 44 | ||
| AK008717.1 | - | 6 | 785 | |||
| AK012091.1 | - | 6 | 788 | |||
| BC024881.1 | - | 6 | 792 | 4022326 | ||
| BC089501.1 | - | 6 | 787 | 6594761 | ||

Zmat2
Genomic Map