siRNA Details
| Gene Symbol: 1700029H14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080781.2 | NP_001074250.1 | 4 | 532 | ||
| NM_025601.3 | NP_079877.1 | 4 | 532 | |||
| GenBank mRNA | AK006340.1 | - | 4 | 477 | ||
| AK006493.1 | - | 4 | 475 | |||
| AK016674.1 | - | 4 | 477 | |||
| BC049575.1 | - | 4 | 498 | 6773883 | ||
| CA464636.1 | - | 4 | 522 | |||

1700029H14Rik
Genomic Map