siRNA Details
| Gene Symbol: 2810004N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025615.2 | NP_079891.2 | 7 | 1337 | ||
| GenBank mRNA | AK012675.1 | - | 7 | 1299 | ||
| AK013989.1 | - | 7 | 1325 | |||
| AK017273.1 | - | 7 | 1297 | |||
| AK143919.1 | - | 1303 | ||||
| AK148731.1 | - | 133 | ||||
| AK166668.1 | - | 7 | 1342 | |||
| BC023675.1 | - | 7 | 1436 | 5354472 | ||

2810004N23Rik
Genomic Map