siRNA Details
| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025626.4 | NP_079902.1 | 4 | 2983 | ||
| GenBank mRNA | AK032875.1 | - | 4 | 2984 | ||
| AK168759.1 | - | 4 | 2949 | |||
| BC021353.1 | - | 4 | 2949 | 5006672 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam107b| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025626.4 | NP_079902.1 | 4 | 2983 | ||
| GenBank mRNA | AK032875.1 | - | 4 | 2984 | ||
| AK168759.1 | - | 4 | 2949 | |||
| BC021353.1 | - | 4 | 2949 | 5006672 | ||