siRNA Details
| Gene Symbol: Aggf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025630.3 | NP_079906.2 | 2 | 605 | ||
| GenBank mRNA | AK008168.1 | - | 2 | 578 | ||
| AK008174.1 | - | 2 | 575 | |||
| AK008399.1 | - | 2 | 578 | |||
| AK017248.1 | - | 2 | 561 | |||
| AK132575.1 | - | 2 | 606 | |||
| AY500995.1 | - | 2 | 578 | |||
| BC052410.1 | - | 2 | 582 | 5716440 | ||

Aggf1
Genomic Map