siRNA Details
| Gene Symbol: Metap1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025633.3 | NP_079909.1 | 6 | 661 | ||
| GenBank mRNA | AJ414378.1 | - | 3 | 524 | ||
| AK005241.1 | - | 6 | 658 | |||
| AK010067.1 | - | 6 | 661 | |||
| AK043123.1 | - | 3 | 1447 | |||
| BC051534.1 | - | 3 | 535 | 3466745 | ||
| BC060739.1 | - | 5 | 2169 | |||

Metap1d
Genomic Map