siRNA Details
| Gene Symbol: Gemin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025656.4 | NP_079932.2 | 10 | 978 | ||
| GenBank mRNA | AK005928.1 | - | 10 | 980 | ||
| AK007515.1 | - | 10 | 979 | |||
| AK013414.1 | - | 10 | 936 | |||
| BC053424.1 | - | 10 | 1407 | 6336522 | ||

Gemin2
Genomic Map