siRNA Details
| Gene Symbol: Ms4a4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025658.4 | NP_079934.2 | 5, 6 | 772 | ||
| GenBank mRNA | AF237915.1 | - | 4, 5 | 446 | ||
| AK003110.1 | - | 5, 6 | 814 | |||
| AK008968.1 | - | 5, 6 | 821 | |||
| BC069842.1 | - | 5, 6 | 772 | 30446709 | ||

Ms4a4d
Genomic Map