siRNA Details
| Gene Symbol: Ribc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025660.2 | NP_079936.1 | 3 | 318 | ||
| GenBank mRNA | AK011592.1 | - | 3 | 318 | ||
| AK017461.1 | - | 3 | 210 | |||
| BC050755.1 | - | 3 | 306 | 6772058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ribc1| Gene Symbol: Ribc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025660.2 | NP_079936.1 | 3 | 318 | ||
| GenBank mRNA | AK011592.1 | - | 3 | 318 | ||
| AK017461.1 | - | 3 | 210 | |||
| BC050755.1 | - | 3 | 306 | 6772058 | ||