siRNA Details
| Gene Symbol: Ubr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025666.2 | NP_079942.1 | 9, 10 | 1175 | ||
| GenBank mRNA | AK088208.1 | - | 9, 10 | 1177 | ||
| AK146308.1 | - | 9, 10 | 1177 | |||
| AK149989.1 | - | 9, 10 | 1130 | |||
| AK155611.1 | - | 9, 10 | 1046 | |||
| BC051678.1 | - | 6, 7 | 703 | |||
| BC058535.1 | - | 8, 9 | 859 | |||
| BC079531.1 | - | 8, 9 | 872 | |||
| BC094323.1 | - | 9, 10 | 1182 | |||
| BC094423.1 | - | 9, 10 | 1133 | 6812389 | ||

Ubr7
Genomic Map