siRNA Details
| Gene Symbol: Tsen15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025677.3 | NP_079953.2 | 5 | 908 | ||
| GenBank mRNA | AK004261.1 | - | 5 | 844 | ||
| AK011608.1 | - | 5 | 865 | |||
| AK017650.1 | - | 5 | 909 | |||
| AK169429.1 | - | 4 | 3663 | |||
| BC024342.1 | - | 5 | 838 | 5002659 | ||

Tsen15
Genomic Map