siRNA Details
| Gene Symbol: Rpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025683.2 | NP_079959.2 | 6 | 644 | ||
| GenBank mRNA | AK007735.1 | - | 6 | 650 | ||
| AK017774.1 | - | 6 | 645 | |||
| AK029088.1 | - | 6 | 643 | |||
| AK088152.1 | - | 6 | 645 | |||
| AK150008.1 | - | 6 | 682 | |||
| BC006953.1 | - | 6 | 638 | |||
| BC019126.1 | - | 6 | 687 | 5008075 | ||

Rpe
Genomic Map