siRNA Details
| Gene Symbol: Nepn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025684.2 | NP_079960.1 | 1, 2 | 220 | ||
| GenBank mRNA | AK017656.1 | - | 1, 2 | 221 | ||
| AK017780.1 | - | 1, 2 | 221 | |||
| AK017842.1 | - | 1, 2 | 92 | |||
| AY938536.1 | - | 1, 2 | 221 | |||
| BC132046.1 | - | 1, 2 | 210 | 40130323 | ||
| BC132048.1 | - | 1, 2 | 210 | 40130325 | ||

Nepn
Genomic Map