siRNA Details
| Gene Symbol: Oser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025699.2 | NP_079975.2 | 4 | 1388 | ||
| GenBank mRNA | AK006969.1 | - | 4 | 1431 | ||
| AK014323.1 | - | 4 | 1407 | |||
| AK088622.1 | - | 4 | 1407 | |||
| AK139692.1 | - | 1560 | ||||
| AK153499.1 | - | 4 | 1425 | |||
| AK208264.1 | - | 1 | 147 | |||
| BC005579.1 | - | 4 | 1388 | 3968876 | ||

Oser1
Genomic Map