siRNA Details
| Gene Symbol: Tmem186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025708.4 | NP_079984.2 | 2 | 1135 | ||
| GenBank mRNA | AK009236.1 | - | 2 | 972 | ||
| AK013147.1 | - | 2 | 964 | |||
| AK014478.1 | - | 2 | 1136 | |||
| AK033686.1 | - | 2 | 973 | |||
| AK088656.1 | - | 2 | 1145 | |||
| AK159727.1 | - | 2 | 1121 | |||
| AK159782.1 | - | 2 | 1121 | |||
| BC027261.1 | - | 2 | 965 | 2649343 | ||
| BC058268.1 | - | 1 | 2197 | |||

Tmem186
Genomic Map