siRNA Details
| Gene Symbol: Snx31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025712.4 | NP_079988.3 | 5 | 450 | ||
| GenBank mRNA | AK014536.1 | - | 5 | 452 | ||
| AK034196.1 | - | 5 | 452 | |||
| AK035226.1 | - | 5 | 451 | |||
| BC061007.1 | - | 5 | 446 | 30314775 | ||

Snx31
Genomic Map