siRNA Details
| Gene Symbol: Ccdc96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025725.2 | NP_080001.1 | 1 | 1002 | ||
| GenBank mRNA | AK014878.1 | - | 1 | 1000 | ||
| AK015646.1 | - | 1 | 1000 | |||
| AK030023.1 | - | 1 | 1002 | |||
| BC125034.1 | - | 1 | 986 | 40096237 | ||

Ccdc96
Genomic Map