siRNA Details
| Gene Symbol: Klhl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025727.3 | NP_080003.1 | 2 | 388 | ||
| GenBank mRNA | AK006288.1 | - | 2 | 375 | ||
| AK014906.1 | - | 2 | 389 | |||
| AK077202.1 | - | 2 | 390 | |||
| AY495337.1 | - | 2 | 389 | |||
| BC064049.1 | - | 2 | 385 | |||
| BC139267.1 | - | 2 | 364 | 8862289 | ||
| BC139268.1 | - | 2 | 364 | 8862290 | ||

Klhl10
Genomic Map