siRNA Details
| Gene Symbol: Kcng4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025734.2 | NP_080010.2 | 3 | 2154 | ||
| GenBank mRNA | AK015001.1 | - | 1 | 620 | ||
| AK015820.1 | - | 1 | 432 | |||
| BC043936.1 | - | 3 | 2154 | 4504272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcng4| Gene Symbol: Kcng4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025734.2 | NP_080010.2 | 3 | 2154 | ||
| GenBank mRNA | AK015001.1 | - | 1 | 620 | ||
| AK015820.1 | - | 1 | 432 | |||
| BC043936.1 | - | 3 | 2154 | 4504272 | ||