siRNA Details
| Gene Symbol: Emc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025736.2 | NP_080012.1 | 6 | 440 | ||
| GenBank mRNA | AF332065.1 | - | 6 | 368 | ||
| AF332066.1 | - | 6 | 368 | |||
| AK014989.1 | - | 6 | 441 | |||
| AK015968.1 | - | 6 | 406 | |||
| AK172887.1 | - | 6 | 385 | |||
| BC004716.1 | - | 6 | 398 | 3582196 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001113785, BC158805, FQ215024, FQ214096.

Emc2
Genomic Map