siRNA Details
| Gene Symbol: Erlec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025745.3 | NP_080021.3 | 7 | 623 | ||
| GenBank mRNA | AK002471.1 | - | 7 | 614 | ||
| AK077120.1 | - | 7 | 616 | |||
| AK133874.1 | - | 7 | 624 | |||
| BC018468.1 | - | 7 | 633 | 3591901 | ||

Erlec1
Genomic Map