siRNA Details
| Gene Symbol: 4933417A18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025750.3 | NP_080026.2 | 3 | 712 | ||
| GenBank mRNA | AK016778.1 | - | 3 | 677 | ||
| AK016838.1 | - | 3 | 713 | |||
| BC100450.1 | - | 3 | 702 | 30919421 | ||
| BC145747.1 | - | 3 | 667 | 40131050 | ||

4933417A18Rik
Genomic Map