siRNA Details
| Gene Symbol: Tmem239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025753.3 | NP_080029.1 | 2 | 917 | ||
| GenBank mRNA | AK016923.1 | - | 2 | 931 | ||
| BC049728.1 | - | 2 | 918 | 6770376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem239| Gene Symbol: Tmem239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025753.3 | NP_080029.1 | 2 | 917 | ||
| GenBank mRNA | AK016923.1 | - | 2 | 931 | ||
| BC049728.1 | - | 2 | 918 | 6770376 | ||