siRNA Details
| Gene Symbol: Gid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025757.4 | NP_080033.3 | 6 | 1301 | ||
| GenBank mRNA | AJ404329.1 | - | 6 | 795 | ||
| AK039435.1 | - | 6 | 1185 | |||
| AK077759.1 | - | 6 | 868 | |||
| AK164644.1 | - | 6 | 1168 | |||
| BC044901.1 | - | 6 | 1003 | 3660276 | ||
| BC046821.1 | - | 6 | 1115 | 6400140 | ||
| BC052418.1 | - | 6 | 1143 | 6401069 | ||

Gid4
Genomic Map