siRNA Details
| Gene Symbol: PGRMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006667.4 | NP_006658.1 | 2 | 523 | ||
| GenBank mRNA | AK313956.1 | - | 2 | 481 | ||
| BC034238.1 | - | 2 | 457 | 3863377 | ||
| CR456993.1 | - | 2 | 404 | |||
| CX762728.1 | - | 2 | 520 | |||
| Y12711.2 | - | 2 | 482 | |||

PGRMC1
Genomic Map