siRNA Details
Gene Symbol: Tmem170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025781.2 | NP_080057.1 | 3 | 1055 | ||
GenBank mRNA | AK018496.1 | - | 3 | 1056 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025781.2 | NP_080057.1 | 3 | 1055 | ||
GenBank mRNA | AK018496.1 | - | 3 | 1056 |