siRNA Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025813.3 | NP_080089.1 | 7 | 656 | ||
| GenBank mRNA | AK004586.1 | - | 7 | 657 | ||
| AK075859.1 | - | 7 | 686 | |||
| AK135235.1 | - | 7 | 657 | |||
| AK136108.1 | - | 7 | 648 | |||
| AK152444.1 | - | 7 | 678 | |||
| AK159554.1 | - | 7 | 684 | |||
| AK218750.1 | - | 5 | 368 | |||
| BC024891.1 | - | 7 | 638 | 3992247 | ||

Mfsd1
Genomic Map